Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Cyanopropyl)dimethylchlorosilane, 94%

Chemical Identifiers

Chemical Name or Material (3-Cyanopropyl)dimethylchlorosilane
CAS 18156-15-5
Molecular Formula C6H12ClNSi
Formula Weight 161.71
MDL Number MFCD00001969
UN Number UN2987
Beilstein 2235062
Synonym Chloro(3-cyanopropyl)dimethylsilane; 4-(Chlorodimethylsilyl)butyronitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0360709

  SDS
alfa aesar™
L0360709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.71
Assay 94%
MDL Number MFCD00001969
CAS 18156-15-5
Beilstein 2235062
Synonym Chloro(3-cyanopropyl)dimethylsilane; 4-(Chlorodimethylsilyl)butyronitrile
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
UN Number UN2987
Quantity 10g
TSCA TSCA
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
Boiling Point 108° to 109°C (15mmHg)
Flash Point 85°C (185°F)
Molecular Formula C6H12ClNSi
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Chemical Name or Material (3-Cyanopropyl)dimethylchlorosilane
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.