Please login to your online account to display your discounted pricing

3-Cyanophenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Cyanophenylboronic acid
CAS 150255-96-2
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H6BNO2
Formula Weight 146.94g/mol
MDL Number MFCD01318967
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359030050

  SDS
thermo fisher scientific
359030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Light orange
Quantity 5g
Chemical Name or Material 3-Cyanophenylboronic acid
CAS 150255-96-2
CAS Min % 96.0
CAS Max % 100.0
Physical Form Light orange powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C7H6BNO2
Formula Weight 146.94g/mol
MDL Number MFCD01318967
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.