Learn More
3-Cyanophenyl isocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 314180050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Cyanophenyl isocyanate | |
16413-26-6 | |
100.0 | |
White to Yellow | |
96% min. (GC) | |
C8H4N2O | |
MFCD00013856 | |
3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038 | |
C1=CC(=CC(=C1)N=C=O)C#N | |
144.13 | |
144.13 | |
Low Melting Solid |
97% | |
96.0 | |
49°C to 54°C | |
Authentic | |
Glass bottle | |
CNC6H4NCO | |
5 g | |
NZHPVPMRNASEQK-UHFFFAOYSA-N | |
3-isocyanatobenzonitrile | |
2733315 | |
97% |
Chemical Identifiers
16413-26-6 | |
144.13 | |
NZHPVPMRNASEQK-UHFFFAOYSA-N | |
2733315 | |
C1=CC(=CC(=C1)N=C=O)C#N |
C8H4N2O | |
MFCD00013856 | |
3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038 | |
3-isocyanatobenzonitrile |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is co
GHS Signal Word: Danger
TSCA : TSCA
RUO – Research Use Only