missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444730010

 View more versions of this product

Catalog No. AC444730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(Cyanomethyl)phenylboronic acid pinacol ester
61°C to 63°C
396131-82-1
C14H18BNO2
LIWPKYSGWSFPEE-UHFFFAOYSA-N
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
2773343
97%
Glass Bottle
1g
97%
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethylphenylboronic acid, pinacol ester, pubchem7790
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CC#N
243.12
243.12

Chemical Identifiers

396131-82-1
243.12
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethylphenylboronic acid, pinacol ester, pubchem7790
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
C14H18BNO2
LIWPKYSGWSFPEE-UHFFFAOYSA-N
2773343
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CC#N
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents