Please login to your online account to display your discounted pricing

3-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Cyanomethyl)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H18BNO2
Formula Weight 243.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444730050

 
thermo fisher scientific
AC444730050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444730010

 
thermo fisher scientific
AC444730010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-(Cyanomethyl)phenylboronic acid pinacol ester
Quantity 5g
CAS 396131-82-1
Assay Percent Range 97%
Molecular Formula C14H18BNO2
Formula Weight 243.11