missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%

$40.39 - $115.64

Chemical Identifiers

CAS 396131-82-1
Molecular Formula C14H18BNO2
Formula Weight 243.11
MDL Number MFCD02179458
Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5261806 SDS Alfa Aesar™
H5261806
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5261803 SDS Alfa Aesar™
H5261803
1g Each for $115.64
AAH52618MD SDS Alfa Aesar™
H52618MD
250mg Each for $40.39
Specifications

Description

Applications
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

396131-82-1
243.11
3-(Cyanomethyl)benzeneboronic acid pinacol ester
C14H18BNO2
MFCD02179458

Specifications

44° to 48°C
396131-82-1
243.11
3-(Cyanomethyl)benzeneboronic acid pinacol ester
5g
C14H18BNO2
MFCD02179458
96%

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger