missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H52618MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
3-(Cyanomethyl)benzeneboronic acid pinacol ester | |
44°C to 48°C | |
MFCD02179458 | |
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester | |
LIWPKYSGWSFPEE-UHFFFAOYSA-N | |
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile | |
2773343 | |
96% |
396131-82-1 | |
C14H18BNO2 | |
250 mg | |
Slightly soluble in water. | |
CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1 | |
243.11 | |
243.11 |
Chemical Identifiers
396131-82-1 | |
243.11 | |
LIWPKYSGWSFPEE-UHFFFAOYSA-N | |
2773343 | |
CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1 |
C14H18BNO2 | |
MFCD02179458 | |
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester | |
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures