Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyanomethyl-4-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Cyanomethyl-4-fluorobenzeneboronic acid pinacol ester
Formula Weight 261.1
Synonym 3-Cyanomethyl-4-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6263203

 
alfa aesar™
H6263203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62632MD

 
alfa aesar™
H62632MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.1
Synonym 3-Cyanomethyl-4-fluorophenylboronic acid pinacol ester
Physical Form Solid
Quantity 1g
Assay 96%
Chemical Name or Material 3-Cyanomethyl-4-fluorobenzeneboronic acid pinacol ester