missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyanobenzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ H54212MD

 View more versions of this product

Catalog No. AAH54212MD



Specifications

Specifications

3-Cyanobenzyl alcohol
250mg
C8H7NO
3-cyanobenzyl alcohol
C1=CC(=CC(=C1)CO)C#N
133.15
133.15
109°C (228°F)
874-97-5
MFCD01321087
PCOFIIVWHXIDGT-UHFFFAOYSA-N
3-(hydroxymethyl)benzonitrile
2734677
97%

Chemical Identifiers

874-97-5
133.15
PCOFIIVWHXIDGT-UHFFFAOYSA-N
2734677
C1=CC(=CC(=C1)CO)C#N
C8H7NO
MFCD01321087
3-cyanobenzyl alcohol
3-(hydroxymethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents