Learn More
3-Cyanobenzyl alcohol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5421206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Cyanobenzyl alcohol | |
109°C (228°F) | |
MFCD01321087 | |
3-cyanobenzyl alcohol, 3-hydroxymethyl benzonitrile, 3-cyanobenzylalcohol, benzonitrile, 3-hydroxymethyl, 3-hyroxymethylbenzonitrile, 3-hydroxymethyl benzenecarbonitrile, m-cyanobenzyl alcohol, 3-cyanophenyl methanol, acmc-209qo5 | |
C1=CC(=CC(=C1)CO)C#N | |
133.15 | |
133.15 |
874-97-5 | |
C8H7NO | |
5 g | |
PCOFIIVWHXIDGT-UHFFFAOYSA-N | |
3-(hydroxymethyl)benzonitrile | |
2734677 | |
97% |
Chemical Identifiers
874-97-5 | |
133.15 | |
PCOFIIVWHXIDGT-UHFFFAOYSA-N | |
2734677 | |
C1=CC(=CC(=C1)CO)C#N |
C8H7NO | |
MFCD01321087 | |
3-cyanobenzyl alcohol, 3-hydroxymethyl benzonitrile, 3-cyanobenzylalcohol, benzonitrile, 3-hydroxymethyl, 3-hyroxymethylbenzonitrile, 3-hydroxymethyl benzenecarbonitrile, m-cyanobenzyl alcohol, 3-cyanophenyl methanol, acmc-209qo5 | |
3-(hydroxymethyl)benzonitrile |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only