Please login to your online account to display your discounted pricing

3-Cyanobenzoyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Cyanobenzoyl chloride
CAS 1711-11-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H4ClNO
Linear Formula NCC6H4COCl
Formula Weight 165.58
MDL Number MFCD00011545
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378130050

  SDS
thermo fisher scientific
AC378130050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC378130010

  SDS
thermo fisher scientific
AC378130010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Formula Weight 165.58
Quantity 5g
Color White to pale yellow or light gray
Melting Point 38° to 44°C
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 Danger
MDL Number MFCD00011545
Molecular Formula C8H4ClNO
CAS 1711-11-1
CAS Min % 97.5
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula NCC6H4COCl
Assay Percent Range 98%
Chemical Name or Material 3-Cyanobenzoyl chloride
Health Hazard 2 GHS P Statement
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.