missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyanobenzoic acid, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC169132500

 View more versions of this product

Catalog No. AC169132500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Cyanobenzoic acid
Glass bottle
217.0°C to 222.0°C
1877-72-1
100.0
C8H5NO2
MFCD00002486
m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
C1=CC(=CC(=C1)C(=O)O)C#N
147.133
147.13
98%
Authentic
White
250g
97.5
98%
NCC6H4CO2H
09, 835
GYLKKXHEIIFTJH-UHFFFAOYSA-N
3-cyanobenzoic acid
15875
Powder

Chemical Identifiers

1877-72-1
147.133
GYLKKXHEIIFTJH-UHFFFAOYSA-N
15875
C1=CC(=CC(=C1)C(=O)O)C#N
C8H5NO2
MFCD00002486
m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
3-cyanobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents