missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyanobenzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H6494806

 View more versions of this product

Catalog No. AAH6494806



Specifications

Specifications

3-Cyanobenzeneboronic acid pinacol ester
78°C to 82°C (decomposition)
214360-46-0
MFCD03789262
FIGQEPXOSAFKTA-UHFFFAOYSA-N
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
2760590
Solid
White
5g
C13H16BNO2
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanobenzeneboronic acid pinacol ester, 3-cyanobenzeneboronic acid, pinacol ester, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, 3-cyanophenylboronicacidpinacolester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem15603
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#N
229.086
229.09
97%

Chemical Identifiers

214360-46-0
229.086
FIGQEPXOSAFKTA-UHFFFAOYSA-N
2760590
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#N
C13H16BNO2
MFCD03789262
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanobenzeneboronic acid pinacol ester, 3-cyanobenzeneboronic acid, pinacol ester, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, 3-cyanophenylboronicacidpinacolester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem15603
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents