Learn More
3-Cyanobenzaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1594303
Description
3-Cyanobenzaldehyde is used in the synthesis of 3-(6,6-dimethyl-5,6-dihydro-4H-benzo[7,8]chromeno[6,5-d]oxazol-2-yl)benzonitrile. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Cyanobenzaldehyde is used in the synthesis of 3-(6,6-dimethyl-5,6-dihydro-4H-benzo[7,8]chromeno[6,5-d]oxazol-2-yl)benzonitrile. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications
3-Cyanobenzaldehyde | |
75°C to 79°C | |
Characteristic | |
MFCD00003344 | |
2205751 | |
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde | |
HGZJJKZPPMFIBU-UHFFFAOYSA-N | |
3-formylbenzonitrile | |
90670 | |
97% |
24964-64-5 | |
209°C to 210°C | |
C8H5NO | |
1 g | |
Air Sensitive | |
Insoluble in water. | |
C1=CC(=CC(=C1)C=O)C#N | |
131.134 | |
131.13 |
Chemical Identifiers
24964-64-5 | |
131.134 | |
HGZJJKZPPMFIBU-UHFFFAOYSA-N | |
90670 | |
C1=CC(=CC(=C1)C=O)C#N |
C8H5NO | |
MFCD00003344 | |
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde | |
3-formylbenzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 246-549-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only