Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-L-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material 3-Cyano-L-phenylalanine
CAS 57213-48-6
Molecular Formula C10H10N2O2
Formula Weight 190.2
MDL Number MFCD01860885
UN Number UN3439
Synonym H-Phe(3-CN)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2747403

 
alfa aesar™
H2747403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27474MD

 
alfa aesar™
H27474MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.2
Chemical Name or Material 3-Cyano-L-phenylalanine
Assay 98%
Molecular Formula C10H10N2O2
MDL Number MFCD01860885
CAS 57213-48-6
UN Number UN3439
Synonym H-Phe(3-CN)-OH
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.