Please login to your online account to display your discounted pricing

3-Cyano-7-ethoxycoumarin, Tocris™

Fluorescent cytochrome P450 substrate

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material 3-Cyano-7-ethoxycoumarin
Assay Percent Range >98%
Molecular Formula C12H9NO3
Formula Weight 215.2
Synonym 7-Ethoxy-2-oxo-2H-1-benzopyran-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

481510

 
r & d systems
RDS481510
10mg Each for $121.23

481550

 
r & d systems
RDS481550
50mg Each for $509.13
Description & Specifications

Specifications

Formula Weight 215.2
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material 3-Cyano-7-ethoxycoumarin
Synonym 7-Ethoxy-2-oxo-2H-1-benzopyran-3-carbonitrile
Molecular Formula C12H9NO3
CAS 117620-77-6
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C