Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-5-(trifluoromethyl)benzeneboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 3-Cyano-5-(trifluoromethyl)benzeneboronic acid
CAS 1212021-62-9
Formula Weight 214.94
MDL Number MFCD03095102
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6126603

 
alfa aesar™
H6126603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61266MD

 
alfa aesar™
H61266MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.94
Chemical Name or Material 3-Cyano-5-(trifluoromethyl)benzeneboronic acid
Physical Form Solid
Assay 97%
MDL Number MFCD03095102
CAS 1212021-62-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
3-Cyano-5-(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.