missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-5-(trifluoromethyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H61266MD

 View more versions of this product

Catalog No. AAH61266MD



Description

Description

Applications
3-Cyano-5-(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

3-Cyano-5-(trifluoromethyl)benzeneboronic acid
1212021-62-9
MFCD03095102
CTJAWCWPRYNDDQ-UHFFFAOYSA-N
[3-cyano-5-(trifluoromethyl)phenyl]boronic acid
56776695
Solid
250mg
C8H5BF3NO2
3-cyano-5-trifluoromethyl phenyl boronic acid
B(C1=CC(=CC(=C1)C(F)(F)F)C#N)(O)O
214.938
214.94
97%

Chemical Identifiers

1212021-62-9
214.938
CTJAWCWPRYNDDQ-UHFFFAOYSA-N
56776695
B(C1=CC(=CC(=C1)C(F)(F)F)C#N)(O)O
C8H5BF3NO2
MFCD03095102
3-cyano-5-trifluoromethyl phenyl boronic acid
[3-cyano-5-(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents