missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-5-methylisoxazole, 97%

$211.27 - $845.08

Chemical Identifiers

CAS 57351-99-2
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.1
MDL Number MFCD09064940
InChI Key JTPIMYCMWLOEDD-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carbonitrile, 5-methyl-3-isoxazolecarbonitrile, 5-methyl-isoxazole-3-carbonitrile, 3-cyano-5-methylisoxazole, 3-cyano-5-methylisoxazole, 3-isoxazolecarbonitrile,5-methyl, 3-isoxazolecarbonitrile, 5-methyl, 5-?methylisoxazole-?3-?carbonitrile
PubChem CID 24229456
IUPAC Name 5-methyl-1,2-oxazole-3-carbonitrile
SMILES CC1=CC(=NO1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6155303 View Documents Alfa Aesar™
H6155303
1g Each for $211.27
 
AAH6155306 View Documents Alfa Aesar™
H6155306
5g Each for $845.08
 
Specifications

Chemical Identifiers

57351-99-2
108.1
JTPIMYCMWLOEDD-UHFFFAOYSA-N
24229456
CC1=CC(=NO1)C#N
C5H4N2O
MFCD09064940
5-methylisoxazole-3-carbonitrile, 5-methyl-3-isoxazolecarbonitrile, 5-methyl-isoxazole-3-carbonitrile, 3-cyano-5-methylisoxazole, 3-cyano-5-methylisoxazole, 3-isoxazolecarbonitrile,5-methyl, 3-isoxazolecarbonitrile, 5-methyl, 5-?methylisoxazole-?3-?carbonitrile
5-methyl-1,2-oxazole-3-carbonitrile

Specifications

1g
C5H4N2O
UN3276
JTPIMYCMWLOEDD-UHFFFAOYSA-N
5-methyl-1,2-oxazole-3-carbonitrile
24229456
97%
57351-99-2
MFCD09064940
5-methylisoxazole-3-carbonitrile, 5-methyl-3-isoxazolecarbonitrile, 5-methyl-isoxazole-3-carbonitrile, 3-cyano-5-methylisoxazole, 3-cyano-5-methylisoxazole, 3-isoxazolecarbonitrile,5-methyl, 3-isoxazolecarbonitrile, 5-methyl, 5-?methylisoxazole-?3-?carbonitrile
CC1=CC(=NO1)C#N
108.1
108.1
3-Cyano-5-methylisoxazole
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.