missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyano-4-methylpyridine 98.0+%, TCI America™

$84.89 - $283.48

Chemical Identifiers

CAS 5444-01-9
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00234272
InChI Key XLAPHZHNODDMDD-UHFFFAOYSA-N
Synonym 3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
PubChem CID 227806
IUPAC Name 4-methylpyridine-3-carbonitrile
SMILES CC1=C(C=NC=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C26081G View Documents Tci America
C26081G
1g Each for $84.89
 
C26085G View Documents Tci America
C26085G
5g Each for $283.48
 
Specifications

Chemical Identifiers

5444-01-9
118.14
XLAPHZHNODDMDD-UHFFFAOYSA-N
227806
CC1=C(C=NC=C1)C#N
C7H6N2
MFCD00234272
3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
4-methylpyridine-3-carbonitrile

Specifications

111°C
47°C
5444-01-9
MFCD00234272
XLAPHZHNODDMDD-UHFFFAOYSA-N
4-methylpyridine-3-carbonitrile
227806
Crystal-Powder at 20°C
3-Cyano-4-methylpyridine
White-Yellow
1g
C7H6N2
3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
CC1=C(C=NC=C1)C#N
118.14
118.14
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No