Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-4-methylpyridine, 97%

$61.03 - $741.32

Chemical Identifiers

Chemical Name or Material 3-Cyano-4-methylpyridine
CAS 5444-01-9
Molecular Formula C7H6N2
Formula Weight 118.14
MDL Number MFCD00234272
Beilstein 112016
Synonym 3-Cyano-4-picoline; 4-Methylnicotinonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1976903

  SDS
alfa aesar™
L1976903
1g Each for $61.03

AAL1976906

  SDS
alfa aesar™
L1976906
5g Each for $188.35

AAL1976914

  SDS
alfa aesar™
L1976914
25g Each for $741.32
Description & Specifications

Specifications

Formula Weight 118.14
Chemical Name or Material 3-Cyano-4-methylpyridine
Health Hazard 1 Warning
MDL Number MFCD00234272
Assay 97%
Beilstein 112016
Molecular Formula C7H6N2
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 5444-01-9
Odor Characteristic
Synonym 3-Cyano-4-picoline; 4-Methylnicotinonitrile
Melting Point 41° to 44°C
Boiling Point 108° to 111°C (20mmHg)
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.