missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-4-methoxy-2-pyridone, 95%

Manufacturer:  Alfa Aesar™ H6427903

 View more versions of this product

Catalog No. AAH6427903



Description

Description

Applications
Used as a intermediate for gimeracil and pharmaceutical application.

Solubility
Soluble in Dimethyl sulfoxide.

Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
Specifications

Specifications

3-Cyano-4-methoxy-2-pyridone
Yellow
1g
C7H6N2O2
UN3439
1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile
MWGIDWPSRDMIQN-UHFFFAOYSA-N
4-methoxy-2-oxo-1H-pyridine-3-carbonitrile
2786702
Crystalline Solid
Hygroscopic
278°C
21642-98-8
MFCD00975442
5929290
Soluble in Dimethyl sulfoxide.
COC1=C(C(=O)NC=C1)C#N
150.137
150.14
95%

Chemical Identifiers

21642-98-8
150.137
MWGIDWPSRDMIQN-UHFFFAOYSA-N
2786702
COC1=C(C(=O)NC=C1)C#N
C7H6N2O2
MFCD00975442
1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile
4-methoxy-2-oxo-1H-pyridine-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents