Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-4-hydroxybenzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 3-Cyano-4-hydroxybenzeneboronic acid pinacol ester
CAS 775351-56-9
Formula Weight 245.09
MDL Number MFCD16994355
Beilstein 10347892
Synonym 2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66434MD

 
alfa aesar™
H66434MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.09
Chemical Name or Material 3-Cyano-4-hydroxybenzeneboronic acid pinacol ester
MDL Number MFCD16994355
Physical Form Solid
Assay 95%
Synonym 2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Quantity 250mg
CAS 775351-56-9
Beilstein 10347892