missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyano-4-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444710010

 View more versions of this product

Catalog No. AC444710010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Cyano-4-fluorophenylboronic acid pinacol ester
75.0°C to 76.0°C
775351-57-0
C13H15BFNO2
RYJOVQGRIOURGT-UHFFFAOYSA-N
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
11402361
97%
Glass Bottle
1g
96% min. (GC)
2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-4-fluorobenzeneboronic acid pinacol ester, 3-cyano-4-fluorophenyl-boronic acid pinacol ester, 3-cyano-4-fluorophenylboronic acid pinacol ester, 3-cyano-4-fluorophenylboronic acid, pinacol ester, benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N
247.08
247.08

Chemical Identifiers

775351-57-0
247.08
2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-4-fluorobenzeneboronic acid pinacol ester, 3-cyano-4-fluorophenyl-boronic acid pinacol ester, 3-cyano-4-fluorophenylboronic acid pinacol ester, 3-cyano-4-fluorophenylboronic acid, pinacol ester, benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
C13H15BFNO2
RYJOVQGRIOURGT-UHFFFAOYSA-N
11402361
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.

GHS P Statement
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a posi

GHS Signal Word: Danger

Documents