Please login to your online account to display your discounted pricing

3-Cyano-4-fluorobenzenesulfonyl chloride, 97%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398780050

 View more versions of this product

Catalog No. AC398780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Quantity 5g
CAS 351003-23-1
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C7H3ClFNO2S
Formula Weight 219.62

Specifications

Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C7H3ClFNO2S
Formula Weight 219.62