Please login to your online account to display your discounted pricing

3-Cyano-4-fluorobenzenesulfonyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C7H3ClFNO2S
Formula Weight 219.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398780010

  SDS
thermo fisher scientific
AC398780010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398780050

  SDS
thermo fisher scientific
AC398780050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.62
Quantity 1g
Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C7H3ClFNO2S
CAS 351003-23-1
Packaging Glass bottle