Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-2-pyridone, 98%

Chemical Identifiers

Chemical Name or Material 3-Cyano-2-pyridone
CAS 20577-27-9
Formula Weight 120.11
MDL Number MFCD00661865
UN Number UN3439
Beilstein 4661670
Synonym 1,2-Dihydro-2-oxo-pyridine-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6425406

  SDS
alfa aesar™
H6425406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6425414

  SDS
alfa aesar™
H6425414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6425422

  SDS
alfa aesar™
H6425422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.11
Melting Point 224° to 226°C
Assay 98%
CAS 20577-27-9
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 1,2-Dihydro-2-oxo-pyridine-3-carbonitrile
Health Hazard 1 Warning
UN Number UN3439
Quantity 5g
Beilstein 4661670
Chemical Name or Material 3-Cyano-2-pyridone
Physical Form Solid
MDL Number MFCD00661865
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.