Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-2-phenoxypyridine, 98%

$108.18 - $1,582.09

Chemical Identifiers

Chemical Name or Material 3-Cyano-2-phenoxypyridine
CAS 14178-15-5
Molecular Formula C12H8N2O
Formula Weight 196.21
MDL Number MFCD00015473
Beilstein 394821
Synonym 2-Phenoxynicotinonitrile; 2-Phenoxypyridine-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1537606

  SDS
alfa aesar™
A1537606
5g Each for $108.18

AAA1537614

  SDS
alfa aesar™
A1537614
25g Each for $467.09

AAA1537622

  SDS
alfa aesar™
A1537622
100g Each for $1,582.09
Description & Specifications

Specifications

Formula Weight 196.21
Melting Point 107° to 109°C
CAS 14178-15-5
Molecular Formula C12H8N2O
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00015473
Assay 98%
Beilstein 394821
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Synonym 2-Phenoxynicotinonitrile; 2-Phenoxypyridine-3-carbonitrile
Chemical Name or Material 3-Cyano-2-phenoxypyridine
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.