missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-fluoropyridine, 98%

CAS: 3939-13-7 | C6H3FN2 | 122.102 g/mol

$60.90 - $183.59

Chemical Identifiers

CAS 3939-13-7
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD03095082
InChI Key USIDQCCXMGJOJM-UHFFFAOYSA-N
Synonym 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
PubChem CID 2783254
IUPAC Name 2-fluoropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)F)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1953803
View Documents
Thermo Scientific Chemicals
L1953803
1 g
Each for $60.90
Only null left
 
AAL1953806
View Documents
Thermo Scientific Chemicals
L1953806
5 g
Each for $183.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3939-13-7
122.102
USIDQCCXMGJOJM-UHFFFAOYSA-N
2783254
C1=CC(=C(N=C1)F)C#N
C6H3FN2
MFCD03095082
3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
2-fluoropyridine-3-carbonitrile

Specifications

3939-13-7
C6H3FN2
1 g
3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
C1=CC(=C(N=C1)F)C#N
122.102
122.1
3-Cyano-2-fluoropyridine
27°C to 30°C
MFCD03095082
386567
USIDQCCXMGJOJM-UHFFFAOYSA-N
2-fluoropyridine-3-carbonitrile
2783254
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only