missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-2,6-dihydroxy-4-(trifluoromethyl)pyridine

$50.15 - $216.97

Chemical Identifiers

CAS 3335-46-4
Molecular Formula C7H3F3N2O2
Molecular Weight (g/mol) 204.108
MDL Number MFCD06200911
InChI Key JRLZWXISQMMITA-UHFFFAOYSA-N
Synonym 2,6-dihydroxy-3-cyano-4-trifluoromethyl pyridine
PubChem CID 2782644
IUPAC Name 2-hydroxy-6-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile
SMILES C1=C(C(=C(NC1=O)O)C#N)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3180703 View Documents Alfa Aesar™
H3180703
1g Each for $50.15
 
AAH3180709 View Documents Alfa Aesar™
H3180709
10g Each for $216.97
 
Specifications

Chemical Identifiers

3335-46-4
204.108
JRLZWXISQMMITA-UHFFFAOYSA-N
2782644
C1=C(C(=C(NC1=O)O)C#N)C(F)(F)F
C7H3F3N2O2
MFCD06200911
2,6-dihydroxy-3-cyano-4-trifluoromethyl pyridine
2-hydroxy-6-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile

Specifications

>220°C
3335-46-4
MFCD06200911
JRLZWXISQMMITA-UHFFFAOYSA-N
2-hydroxy-6-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile
2782644
3-Cyano-2,6-dihydroxy-4-(trifluoromethyl)pyridine
1g
C7H3F3N2O2
2,6-dihydroxy-3-cyano-4-trifluoromethyl pyridine
C1=C(C(=C(NC1=O)O)C#N)C(F)(F)F
204.108
204.11
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures