Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyano-2-(2-formyl-1-pyrrolyl)-4-methoxypyridine, 97%

$348.23 - $348.23

Chemical Identifiers

Chemical Name or Material 3-Cyano-2-(2-formyl-1-pyrrolyl)-4-methoxypyridine
CAS 338412-33-2
Molecular Formula C12H9N3O2
Formula Weight 227.22g/mol
MDL Number MFCD00975451
Synonym 2-(2-Formyl-1-pyrrolyl)-4-methoxynicotinonitrile; 2-(2-Formyl-1-pyrrolyl)-4-methoxypyridine-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5045803

 
alfa aesar™
H50458-03
1g Each for $348.23
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 3-Cyano-2-(2-formyl-1-pyrrolyl)-4-methoxypyridine
CAS 338412-33-2
Assay 97%
Molecular Formula C12H9N3O2
Formula Weight 227.22g/mol
MDL Number MFCD00975451
Synonym 2-(2-Formyl-1-pyrrolyl)-4-methoxynicotinonitrile; 2-(2-Formyl-1-pyrrolyl)-4-methoxypyridine-2-carbonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.