Please login to your online account to display your discounted pricing

3-CPMT, Tocris


Dopamine uptake inhibitor

Quantity 10mg

Manufacturer: r & d systems  091710

 View more versions of this product

Catalog No. 091710

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 3-CPMT
Quantity 10mg
CAS 14008-79-8
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C21H24ClNO·HCl
Formula Weight 378.34
Synonym 3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride

Specifications

Chemical Name or Material 3-CPMT
Assay Percent Range >99%
Molecular Formula C21H24ClNO·HCl
Formula Weight 378.34
Synonym 3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
Accessories

Store at Room Temperature