Please login to your online account to display your discounted pricing

3-CPMT, Tocris

Dopamine uptake inhibitor

$88.05 - $376.43

Chemical Identifiers

Chemical Name or Material 3-CPMT
Assay Percent Range >99%
Molecular Formula C21H24ClNO·HCl
Formula Weight 378.34
Synonym 3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

091710

 
r & d systems
091710
10mg Each for $88.05

091750

 
r & d systems
091750
50mg Each for $376.43
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 378.34
Chemical Name or Material 3-CPMT
CAS 14008-79-8
Assay Percent Range >99%
Molecular Formula C21H24ClNO·HCl
Synonym 3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature