Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorotoluene, 98%

Chemical Identifiers

Chemical Name or Material 3-Chlorotoluene
CAS 108-41-8
Molecular Formula C7H7Cl
Formula Weight 126.59
MDL Number MFCD00000595
UN Number UN2238
Beilstein 1903632
Merck Index 14,2171
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA169910E

  SDS
alfa aesar™
A169910E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1699122

  SDS
alfa aesar™
A1699122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1699136

  SDS
alfa aesar™
A1699136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.59
Chemical Name or Material 3-Chlorotoluene
TSCA TSCA
Assay 98%
Molecular Formula C7H7Cl
MDL Number MFCD00000595
CAS 108-41-8
UN Number UN2238
Beilstein 1903632
Merck Index 14,2171
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROTOLUENES
Flash Point 50°C (122°F)
Boiling Point 160° to 162°C
Melting Point −48°C
Quantity 2500g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.