Please login to your online account to display your discounted pricing

3-Chlorothiophenol, 97%, ACROS Organics™


Quantity 1mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  222000010

 View more versions of this product

Catalog No. AC222000010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 144.62
Quantity 1mL
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 218-010-0
Beilstein 06, 326
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.6010 to 1.6040 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Name Note 97%
Infrared Spectrum Authentic
MDL Number MFCD00004839
Assay Percent Range 97%
Linear Formula ClC6H4SH
CAS 2037-31-2
Molecular Formula C6H5ClS
Chemical Name or Material 3-Chlorothiophenol
Synonym 3-Chlorobenzenethiol
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 3-Chlorothiophenol
Name Note 97%
CAS 2037-31-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H5ClS
Linear Formula ClC6H4SH
Formula Weight 144.62
MDL Number MFCD00004839
Synonym 3-Chlorobenzenethiol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents