Learn More
3-Chlorothiophene, 98%, may contain up to 2% DMF, Thermo Scientific Chemicals
$84.77 - $787.49
Chemical Identifiers
CAS | 17249-80-8 |
---|---|
Molecular Formula | C4H3ClS |
Molecular Weight (g/mol) | 118.578 |
MDL Number | MFCD00043887 |
InChI Key | QUBJDMPBDURTJT-UHFFFAOYSA-N |
Synonym | thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h |
PubChem CID | 87017 |
IUPAC Name | 3-chlorothiophene |
SMILES | C1=CSC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2266906
|
Thermo Scientific Chemicals
B2266906 |
5 g |
Each for $84.77
|
|
|||||
AAB2266914
|
Thermo Scientific Chemicals
B2266914 |
25 g |
Each for $255.67
|
|
|||||
AAB2266922
|
Thermo Scientific Chemicals
B2266922 |
100 g |
Each for $787.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17249-80-8 | |
118.578 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
87017 | |
C1=CSC=C1Cl |
C4H3ClS | |
MFCD00043887 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
3-chlorothiophene |
Specifications
17249-80-8 | |
137°C to 139°C | |
Unpleasant | |
1.552 | |
5 g | |
105255 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
3-chlorothiophene | |
87017 | |
98% | |
may contain up to 2% DMF |
1.276 | |
37°C (98°F) | |
C4H3ClS | |
MFCD00043887 | |
UN1992 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
C1=CSC=C1Cl | |
118.578 | |
118.59 | |
3-Chlorothiophene |
Safety and Handling
GHS H Statement
H331-H360-H318-H302-H312-H315-H335
Toxic if inhaled.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H318-H335-H350-H360D-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only