missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chlorothiophene, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC258800250
Specifications
3-Chlorothiophene | |
37°C | |
Glass bottle | |
1.276 | |
137.0°C to 139.0°C | |
25g | |
17249-80-8 | |
100.0 | |
C4H3ClS | |
3-chloranylthiophene, 3-chloro thiophene, 3-chloro-thiophene, 3-chlorothiophene, 3-chlorthiophen, 3-thienyl chloride, acmc-1cdmv, ksc175g2h, pubchem5477, thiophene, 3-chloro | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
3-chlorothiophene | |
87017 | |
Liquid |
1.2760g/mL | |
Authentic | |
1.5520 to 1.5540 | |
0.1% max. (K.F.) | |
Colorless to Yellow | |
98% | |
97.5 | |
95% min. (GC) | |
MFCD00043887 | |
Solubility in water: <0.2g/L (20°C). | |
C1=CSC=C1Cl | |
118.59 | |
118.59 | |
96% |
Chemical Identifiers
17249-80-8 | |
118.59 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
87017 | |
C1=CSC=C1Cl |
C4H3ClS | |
MFCD00043887 | |
3-chloranylthiophene, 3-chloro thiophene, 3-chloro-thiophene, 3-chlorothiophene, 3-chlorthiophen, 3-thienyl chloride, acmc-1cdmv, ksc175g2h, pubchem5477, thiophene, 3-chloro | |
3-chlorothiophene |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Flammable liquid and vapor.
May damage the unborn child.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Danger
TSCA : TSCA
Recommended Storage : Product darkens on storage