missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorothiophene, 96%, ACROS Organics™

$64.54 - $64.54

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.59
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym 3-chloranylthiophene, 3-chloro thiophene, 3-chloro-thiophene, 3-chlorothiophene, 3-chlorthiophen, 3-thienyl chloride, acmc-1cdmv, ksc175g2h, pubchem5477, thiophene, 3-chloro
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC258800050
SDS View Documents
Thermo Fisher Scientific
AC258800050
5g Glass bottle
Each for $64.54
 
AC258800250
SDS View Documents
Thermo Fisher Scientific
AC258800250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

17249-80-8
118.59
QUBJDMPBDURTJT-UHFFFAOYSA-N
87017
C1=CSC=C1Cl
C4H3ClS
MFCD00043887
3-chloranylthiophene, 3-chloro thiophene, 3-chloro-thiophene, 3-chlorothiophene, 3-chlorthiophen, 3-thienyl chloride, acmc-1cdmv, ksc175g2h, pubchem5477, thiophene, 3-chloro
3-chlorothiophene

Specifications

1.2760g/mL
Authentic
1.5520 to 1.5540
0.1% max. (K.F.)
Colorless to Yellow
17249-80-8
100.0
C4H3ClS
3-chloranylthiophene, 3-chloro thiophene, 3-chloro-thiophene, 3-chlorothiophene, 3-chlorthiophen, 3-thienyl chloride, acmc-1cdmv, ksc175g2h, pubchem5477, thiophene, 3-chloro
QUBJDMPBDURTJT-UHFFFAOYSA-N
3-chlorothiophene
87017
Liquid
3-Chlorothiophene, 98%
37°C
Glass bottle
1.276
137.0°C to 139.0°C
5g
97.5
95% min. (GC)
MFCD00043887
Solubility in water: <0.2g/L (20°C).
C1=CSC=C1Cl
118.59
118.59
96%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Flammable liquid and vapor.
May damage the unborn child.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Danger

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS