Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorothiobenzamide, 97%

$100.96 - $304.36

Chemical Identifiers

Chemical Name or Material 3-Chlorothiobenzamide
CAS 2548-79-0
Molecular Formula C7H6ClNS
Formula Weight 171.65
MDL Number MFCD00276993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2642303

  SDS
alfa aesar™
H2642303
1g Each for $100.96

AAH2642306

  SDS
alfa aesar™
H2642306
5g Each for $304.36
Description & Specifications

Specifications

Formula Weight 171.65
Molecular Formula C7H6ClNS
Health Hazard 1 Warning
Melting Point 114° to 118°C
Assay 97%
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2548-79-0
Quantity 1g
Chemical Name or Material 3-Chlorothiobenzamide
MDL Number MFCD00276993
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.