Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorothioanisole, 99%

$43.55 - $150.85

Chemical Identifiers

Chemical Name or Material 3-Chlorothioanisole
CAS 4867-37-2
Molecular Formula C7H7ClS
Formula Weight 158.65
MDL Number MFCD00040127
Beilstein 1931485
Synonym 3-Chlorophenyl methyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1134603

  SDS
alfa aesar™
L1134603
1g Each for $43.55

AAL1134606

  SDS
alfa aesar™
L1134606
5g Each for $150.85
Description & Specifications

Specifications

Formula Weight 158.65
Chemical Name or Material 3-Chlorothioanisole
Molecular Formula C7H7ClS
MDL Number MFCD00040127
CAS 4867-37-2
Synonym 3-Chlorophenyl methyl sulfide
Odor Unpleasant
Assay 99%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P261-P280f
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
Flash Point 88°C (190°F)
Boiling Point 218°C
Quantity 1g
Beilstein 1931485
Safety and Handling

Warning

GHS P Statement
P261-P280f
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.