missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorothioanisole, 99%

Manufacturer:  Alfa Aesar™ L1134606

 View more versions of this product

Catalog No. AAL1134606



Specifications

Specifications

3-Chlorothioanisole
Unpleasant
5g
C7H7ClS
1931485
PTGSDZVASWKUHK-UHFFFAOYSA-N
1-chloro-3-methylsulfanylbenzene
97585
99%
88°C (190°F)
218°C
4867-37-2
MFCD00040127
3-chlorothioanisole, 3-chlorophenyl methyl sulfide, 3-chloro thioanisole, 1-chloro-3-methylsulfanyl benzene, m-chlorothioanisole, 3-methylthio chlorobenzene, 3-cpms, 3-chlorothioanisol, pubchem10644, acmc-1bnc1
CSC1=CC(=CC=C1)Cl
158.643
158.65

Chemical Identifiers

4867-37-2
158.643
PTGSDZVASWKUHK-UHFFFAOYSA-N
97585
CSC1=CC(=CC=C1)Cl
C7H7ClS
MFCD00040127
3-chlorothioanisole, 3-chlorophenyl methyl sulfide, 3-chloro thioanisole, 1-chloro-3-methylsulfanyl benzene, m-chlorothioanisole, 3-methylthio chlorobenzene, 3-cpms, 3-chlorothioanisol, pubchem10644, acmc-1bnc1
1-chloro-3-methylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

GHS P Statement
P261-P280f
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.

Warning

Documents