Learn More
3-Chloropyridine-4-carboxaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55852MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Chloropyridine-4-carboxaldehyde | |
58°C to 62°C | |
MFCD06200712 | |
Air Sensitive | |
ZVGDKOQPJCOCLI-UHFFFAOYSA-N | |
3-chloropyridine-4-carbaldehyde | |
2762995 | |
98% |
72990-37-5 | |
C6H4ClNO | |
250 mg | |
3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f | |
C1=CN=CC(=C1C=O)Cl | |
141.554 | |
141.56 |
Chemical Identifiers
72990-37-5 | |
141.554 | |
ZVGDKOQPJCOCLI-UHFFFAOYSA-N | |
2762995 | |
C1=CN=CC(=C1C=O)Cl |
C6H4ClNO | |
MFCD06200712 | |
3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f | |
3-chloropyridine-4-carbaldehyde |
Safety and Handling
GHS H Statement
H301-H315-H319-H317
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only