missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloropyridine-4-carboxaldehyde, 98%

Manufacturer:  Alfa Aesar™ H5585203

 View more versions of this product

Catalog No. AAH5585203



Specifications

Specifications

3-Chloropyridine-4-carboxaldehyde
1g
C6H4ClNO
3-chloro-4-formylpyridine
C1=CN=CC(=C1C=O)Cl
141.554
141.56
58°C to 62°C
72990-37-5
MFCD06200712
ZVGDKOQPJCOCLI-UHFFFAOYSA-N
3-chloropyridine-4-carbaldehyde
2762995
98%

Chemical Identifiers

72990-37-5
141.554
ZVGDKOQPJCOCLI-UHFFFAOYSA-N
2762995
C1=CN=CC(=C1C=O)Cl
C6H4ClNO
MFCD06200712
3-chloro-4-formylpyridine
3-chloropyridine-4-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H317
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents