Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloropyridine-2-carboxylic acid, 97%

$227.81 - $828.14

Chemical Identifiers

Chemical Name or Material 3-Chloropyridine-2-carboxylic acid
CAS 57266-69-0
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD01646325
Synonym 3-Chloropicolinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2602403

 
alfa aesar™
H2602403
1g Each for $828.14

AAH26024MD

 
alfa aesar™
H26024MD
250mg Each for $227.81
Description & Specifications

Specifications

Formula Weight 157.56
Chemical Name or Material 3-Chloropyridine-2-carboxylic acid
Assay 97%
Molecular Formula C6H4ClNO2
MDL Number MFCD01646325
CAS 57266-69-0
Synonym 3-Chloropicolinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 131°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.