missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropyrazine-2-carboxamide, TCI America™

Manufacturer:  TCI America C33761G

 View more versions of this product

Catalog No. C33761G



Specifications

Specifications

3-Chloropyrazine-2-carboxamide
189°C
21279-62-9
MFCD09863915
C1=CN=C(C(=N1)C(=O)N)Cl
157.557
157.56
≥98.0% (HPLC,N)
White-Yellow
1g
C5H4ClN3O
YHPMRHPLAQSPHJ-UHFFFAOYSA-N
3-chloropyrazine-2-carboxamide
301266
Crystal-Powder at 20°C

Chemical Identifiers

21279-62-9
157.557
YHPMRHPLAQSPHJ-UHFFFAOYSA-N
3-chloropyrazine-2-carboxamide
C5H4ClN3O
MFCD09863915
301266
C1=CN=C(C(=N1)C(=O)N)Cl
Safety and Handling

Safety and Handling

TSCA : No

Documents