missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropyldimethoxymethylsilane, TCI America™

$50.42 - $165.00

Chemical Identifiers

CAS 18171-19-2
Molecular Formula C6H15ClO2Si
Molecular Weight (g/mol) 182.719
MDL Number MFCD00053912
InChI Key KNTKCYKJRSMRMZ-UHFFFAOYSA-N
Synonym 3-(Methyldimethoxysilyl)propyl Chloride
PubChem CID 87491
IUPAC Name 3-chloropropyl-dimethoxy-methylsilane
SMILES CO[Si](C)(CCCCl)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C11685G View Documents TCI America
C11685G
5g Each for $50.42
 
C116825G View Documents TCI America
C116825G
25g Each for $165.00
 
Specifications

Chemical Identifiers

18171-19-2
182.719
KNTKCYKJRSMRMZ-UHFFFAOYSA-N
87491
CO[Si](C)(CCCCl)OC
C6H15ClO2Si
MFCD00053912
3-(Methyldimethoxysilyl)propyl Chloride
3-chloropropyl-dimethoxy-methylsilane

Specifications

185°C
5g
C6H15ClO2Si
1993
KNTKCYKJRSMRMZ-UHFFFAOYSA-N
3-chloropropyl-dimethoxy-methylsilane
87491
Clear Liquid at 20°C
3-Chloropropyldimethoxymethylsilane
Yellow
18171-19-2
MFCD00053912
3-(Methyldimethoxysilyl)propyl Chloride
CO[Si](C)(CCCCl)OC
182.719
182.72
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2078

TSCA : Yes