Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Chloropropyl)trimethylsilane, 98%

Chemical Identifiers

Chemical Name or Material (3-Chloropropyl)trimethylsilane
CAS 2344-83-4
Molecular Formula ClCH2CH2CH2Si(CH3)3
Formula Weight 150.73
MDL Number MFCD00045299
UN Number UN1993
Beilstein 1732625
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0445806

 
alfa aesar™
L0445806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0445814

 
alfa aesar™
L0445814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.73
Chemical Name or Material (3-Chloropropyl)trimethylsilane
TSCA TSCA
Assay 98%
Molecular Formula ClCH2CH2CH2Si(CH3)3
MDL Number MFCD00045299
CAS 2344-83-4
Beilstein 1732625
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1993
Boiling Point 150° to 152°C
Quantity 5g
Flash Point 49°C (120°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Applications
(3-Chloropropyl)trimethylsilane used in the preparation of: 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions, 5-trimethylsilyl pentanol, N-(tri-methylsilylalkyl)diamines.

Solubility
Not miscible in water.

Notes
Store under inert gas. Moisture sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.