Learn More
(3-Chloropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2251736
Description
(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
195°C to 197°C | |
-50°C | |
500 g | |
MFCD00000997 | |
78°C (172°F) | |
Hydrolyzes in water. | |
CO[Si](CCCCl)(OC)OC | |
198.718 | |
198.72 | |
Recognizable | |
Moisture sensitive |
(3-Chloropropyl)trimethoxysilane | |
2530-87-2 | |
C6H15ClO3Si | |
969627 | |
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
3-chloropropyl(trimethoxy)silane | |
62449 | |
97% | |
1.42 | |
1.077 |
Chemical Identifiers
2530-87-2 | |
198.718 | |
OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
62449 | |
CO[Si](CCCCl)(OC)OC |
C6H15ClO3Si | |
MFCD00000997 | |
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
3-chloropropyl(trimethoxy)silane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 219-787-9
RTECSNumber : VV2680000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only