Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Chloropropyl)trichlorosilane, 97%

$84.82 - $84.82

Chemical Identifiers

Chemical Name or Material (3-Chloropropyl)trichlorosilane
CAS 2550-06-3
Molecular Formula C3H6Cl4Si
Linear Formula Cl(CH2)3SiCl3
Formula Weight 211.98
MDL Number MFCD00013694
UN Number UN2987
Beilstein 1737851
Synonym Trichloro(3-chloropropyl)silane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1777022

  SDS
alfa aesar™
A1777022
100g Each for $84.82

AAA1777014

  SDS
alfa aesar™
A1777014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.98
Boiling Point 181° to 183°C
MDL Number MFCD00013694
Chemical Name or Material (3-Chloropropyl)trichlorosilane
Flash Point 84°C (183°F)
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Quantity 100g
Odor Acrid
TSCA TSCA
Assay 97%
Beilstein 1737851
Synonym Trichloro(3-chloropropyl)silane
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 1 Danger
Molecular Formula C3H6Cl4Si
UN Number UN2987
Linear Formula Cl(CH2)3SiCl3
CAS 2550-06-3
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.