Please login to your online account to display your discounted pricing

3-Chloropropyl isocyanate, 96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314590010

 View more versions of this product

Catalog No. AC314590010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 119.55
Quantity 1g
Color Clear colorless
TSCA TSCA
Density 1.16g/mL
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Infrared Spectrum Authentic
Molecular Formula C4H6ClNO
MDL Number MFCD00010384
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Chloropropyl isocyanate
Name Note 96%
Assay Percent Range 96%
CAS 13010-19-0
Refractive Index 1.4490 to 1.4530 (20°C, 589nm)
Linear Formula Cl(CH)3NCO
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 3-Chloropropyl isocyanate
Name Note 96%
CAS 13010-19-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C4H6ClNO
Linear Formula Cl(CH)3NCO
Formula Weight 119.55
MDL Number MFCD00010384
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.

SDS & Safety Documents